| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
TC01002432.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
ENST00000373795.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
ENST00000434636.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
ENST00000466448.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
ENST00000546138.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037447.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200209.hg.1 |
TC0100013497.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000373795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000466448.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000477193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000489898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000497015.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000605204.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
ENST00000622143.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013497.hg.1 |
NM_005626.4 |
Refseq |