| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
ENST00000370812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
ENST00000370813.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
ENST00000445065.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
ENST00000487906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043548.hg.1 |
TC01002796.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
ENST00000370812.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
ENST00000370813.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
ENST00000445065.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
ENST00000487906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01043546.hg.1 |
TC01002796.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000478391.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100014638.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209183.hg.1 |
TC0100014638.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000359130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000370812.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000445065.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
ENST00000487906.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
NM_005482.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100209185.hg.1 |
TC0100014638.hg.1 |
Clariom D TC |