| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8063390 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8063391 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8063393 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8063392 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8063389 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1556896_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005733.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005733.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005733.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005732.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005732.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005732.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005728.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005728.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005728.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005730.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005730.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005730.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005731.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005731.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005731.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005727.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005727.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005727.hg.1 |
TC20000402.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007687.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153614.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153599.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153610.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153612.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153613.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153600.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153607.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
ENST00000457853.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153608.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153601.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
ENST00000371639.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
NR_034124.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153604.hg.1 |
TC2000007687.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
1556896_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556896_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556896_at7 |
- |
- |
| Human Genome U95 Set |
60344_PM_at |
- |
- |
| Human Genome U95 Set |
60344_at |
- |
- |