| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000355968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000395070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000451562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000468812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000480603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
ENST00000489459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
NM_021130.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004388.hg.1 |
TC07000278.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000244636.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000355968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000395070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000451562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000468812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000480603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
ENST00000489459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
NM_021130.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004390.hg.1 |
TC07000278.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008469.hg.1 |
ENST00000438507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008469.hg.1 |
ENST00000438507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010360.hg.1 |
ENST00000414193.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000244636.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000355968.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000395070.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000451562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000468812.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000480603.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
ENST00000489459.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
NM_021130.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07004392.hg.1 |
TC07000278.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010360.hg.1 |
ENST00000414193.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007213.hg.1 |
ENST00000414949.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008469.hg.1 |
ENST00000438507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008469.hg.1 |
ENST00000438507.1 |
Ensembl |