Search
Search

KIAA1958| Gene Symbol: KIAA1958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287036.1 | NP_001273965.1 | 5 | 4666 | 87 | |
| NM_001287038.1 | NP_001273967.1 | 4 | 4572 | 87 | ||
| NM_133465.3 | NP_597722.1 | 4 | 4582 | 87 | ||
| GenBank mRNA | AB075838.1 | - | 3 | 4082 | 87 | |
Set Membership: |
|