| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000527086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528207.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000528690.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000532766.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012218.hg.1 |
ENST00000569326.1 |
Ensembl |