Search
Search

KIAA1586| Gene Symbol: KIAA1586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286275.1 | NP_001273204.1 | 3 - 4 | 390 | 129 | |
| NM_001286276.1 | NP_001273205.1 | 3 - 4 | 342 | 129 | ||
| NM_020931.3 | NP_065982.1 | 3 - 4 | 412 | 129 | ||
| XM_011514767.2 | XP_011513069.1 | 3 - 4 | 253 | 129 | ||
| GenBank mRNA | AK290988.1 | - | 3 - 4 | 206 | 129 | |
| BC029372.1 | - | 3 - 4 | 221 | 129 | ||
| BC041125.1 | - | 3 - 4 | 358 | 5785266 | 129 | |
| BF699031.1 | - | 3 - 4 | 341 | 128 | ||
| BG028983.1 | - | 178 | 129 | |||
Set Membership: |
|