Search
Search

SCGB2B2| Gene Symbol: SCGB2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025591.4 | NP_001020762.1 | 2 - 3 | 3628 | 78 | |
| GenBank mRNA | AK093495.1 | - | 2 - 3 | 2154 | 78 | |
| BC093909.1 | - | 1 - 2 | 89 | 78 | ||
| BC093935.1 | - | 1 - 2 | 89 | 78 | ||
Set Membership: |
|