Search
Search
| Gene Symbol: NRTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004558.3 | NP_004549.1 | 1 - 2 | 525 | 93 | |
| XM_011528041.2 | XP_011526343.1 | 2 - 3 | 629 | 93 | ||
| GenBank mRNA | AL161995.1 | - | 1 - 2 | 525 | 93 | |
| BC137399.1 | - | 1 - 2 | 278 | 9021397 | 93 | |
| BC137400.1 | - | 1 - 2 | 278 | 9021398 | 93 | |
| U78110.1 | - | 1 - 2 | 166 | 93 | ||
Set Membership: |
|