Search
Search

CCDC190| Gene Symbol: CCDC190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178550.4 | NP_848645.3 | 3 - 4 | 490 | 61 | |
| XM_005245125.3 | XP_005245182.1 | 3 - 4 | 3108 | 61 | ||
| XM_006711289.3 | XP_006711352.1 | 3 - 4 | 590 | 61 | ||
| XM_011509477.2 | XP_011507779.1 | 3 - 4 | 3111 | 61 | ||
| GenBank mRNA | AK123573.1 | - | 3 - 4 | 487 | 61 | |
Set Membership: |
|