| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2010912 |
L40172.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2010912 |
L33768.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2010912 |
BC131646.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2010912 |
BC131647.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2010912 |
BC105577.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
317395 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437030 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
547418 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
AF323659.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
AK037241.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
BC119471.2 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
NM_009593.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
U34920.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532780 |
Z48745.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828069 |
AK044495.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
630657 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
459116 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75182 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
155072 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155415 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169228 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2010912 |
L40172.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2010912 |
L33768.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2010912 |
BC131646.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2010912 |
BC131647.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2010912 |
BC105577.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P288406 |
L32955.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P288406 |
BC105577.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P288406 |
S82815.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P512847 |
AK088365.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25958248 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25958245 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25958250 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25958252 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25958242 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155415 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169228 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06551714 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06551689 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
BC105577.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
AK088365.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
AK038268.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
NM_010589.6 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P355360 |
NM_013564.6 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P512847 |
AK088365.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109865 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155415 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169228 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07005926 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07005930 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958242 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958330 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958313 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958314 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958225 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958359 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958392 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958203 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958317 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958296 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958355 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958345 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958353 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958293 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958297 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958233 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958303 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958358 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958301 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958361 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958344 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958302 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958328 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958238 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958332 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958295 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958321 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958396 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958211 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958323 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958229 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958312 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958310 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958299 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958241 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958316 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958368 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958245 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958337 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958324 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958306 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958403 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958346 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958187 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958308 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958349 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958386 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25958221 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958245 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958219 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958221 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07005936 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958240 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958237 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958233 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07005930 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958248 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958235 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958230 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958227 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958249 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958222 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958232 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07005928 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958225 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25958223 |
- |
- |