| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400014971.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014971.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014971.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014972.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014972.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014972.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014978.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014978.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014978.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014984.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014984.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014984.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014974.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014974.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014974.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014966.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014966.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014966.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014970.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014970.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014970.mm.1 |
TC1400001993.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400014979.mm.1 |
NM_133676.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014979.mm.1 |
NM_133676.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400014979.mm.1 |
TC1400001993.mm.1 |
MTA TC |