| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077765 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077768 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077769 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077767 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077766 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
632375 |
NM_146732.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
695791 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
650698 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149219 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P445574 |
AF462069.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077758 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077779 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077749 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077765 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077752 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077744 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077775 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077767 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077772 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077789 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077759 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077764 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077738 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077753 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077783 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077770 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077747 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077792 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077778 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077754 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077741 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077781 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077760 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077750 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077786 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077763 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077769 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077756 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077784 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28077793 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149219 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P445574 |
AF462069.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
BC057990.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
AK075765.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
BC036161.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
AJ489814.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P487436 |
NM_145829.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108029 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149219 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735613 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735622 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735609 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735595 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735608 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735602 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09735612 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077767 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077768 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077769 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077766 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28077765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077772 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077789 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077783 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077823 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077779 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077784 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077790 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077837 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077793 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077773 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077764 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077835 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077849 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077808 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077758 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077829 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077830 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077825 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077781 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077810 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077792 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077767 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077761 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077845 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077814 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28077794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077741 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077779 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573069 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077793 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077747 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077764 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09349800 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09735589 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09136551 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09101375 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077744 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077758 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077749 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077769 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077756 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077790 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077738 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077745 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077727 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077750 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077733 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573072 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077789 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077796 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077735 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09349798 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09077874 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077802 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077786 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077792 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077730 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077783 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09331456 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077798 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077799 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077775 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077773 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077770 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077752 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077777 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077724 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077757 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09735599 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077761 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573070 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077763 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077781 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28077772 |
- |
- |