Search
Search
| Gene Symbol: Fis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025562.3 | NP_079838.1 | 1 - 2 | 146 | 68 | |
| GenBank mRNA | AK004479.1 | - | 1 - 2 | 118 | 68 | |
| AK004917.1 | - | 1 - 2 | 100 | 68 | ||
| AK008078.1 | - | 1 - 2 | 138 | 68 | ||
| AK008323.1 | - | 1 - 2 | 123 | 68 | ||
| AK008916.1 | - | 1 - 2 | 100 | 68 | ||
| AW988976.1 | - | 1 - 2 | 146 | 68 | ||
| BC010783.1 | - | 1 - 2 | 119 | 4221162 | 68 | |
Set Membership: |
|