Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009180.3 | NP_033206.2 | 7 - 8 | 953 | 68 | |
GenBank mRNA | AK004613.1 | - | 7 - 8 | 954 | 68 | |
AK136381.1 | - | 956 | 68 | |||
BC003276.1 | - | 7 - 8 | 1855 | 68 | ||
BC010208.1 | - | 905 | 68 | |||
BC023076.1 | - | 1 - 2 | 1401 | 68 | ||
EU937514.1 | - | 1 - 2 | 1400 | 68 | ||
X93999.1 | - | 7 - 8 | 917 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |