Search
Search
| Gene Symbol: Rtkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136227.1 | NP_001129699.1 | 2 - 3 | 454 | 71 | |
| NM_009106.2 | NP_033132.2 | 3 - 4 | 861 | 71 | ||
| NM_133641.2 | NP_598396.2 | 2 - 3 | 1365 | 71 | ||
| GenBank mRNA | AK076155.1 | - | 2 - 3 | 375 | 71 | |
| BC004824.1 | - | 2 - 3 | 695 | 71 | ||
| BC013820.1 | - | 2 - 3 | 1339 | 3981927 | 71 | |
| U54638.1 | - | 3 - 4 | 864 | 71 | ||
Set Membership: |
|