Search
Search

Fam173a| Gene Symbol: Fam173a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285982.1 | NP_001272911.1 | 2 - 3 | 164 | 64 | |
| NM_001285984.1 | NP_001272913.1 | 2 - 3 | 164 | 64 | ||
| NM_145410.4 | NP_663385.2 | 2 - 3 | 449 | 64 | ||
| GenBank mRNA | AK132031.1 | - | 2 - 3 | 323 | 64 | |
| AK146214.1 | - | 2 - 3 | 430 | 64 | ||
| AK162337.1 | - | 2 - 3 | 425 | 64 | ||
| BB855744.1 | - | 2 - 3 | 450 | 64 | ||
| BC008155.1 | - | 2 - 3 | 173 | 3154632 | 64 | |
| BC096050.1 | - | 2 - 3 | 344 | 5119802 | 64 | |
Set Membership: |
|