Search
Search

Nup214| Gene Symbol: Nup214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172268.2 | NP_758472.2 | 14 - 15 | 2134 | 58 | |
| GenBank mRNA | AK122189.1 | - | 14 - 15 | 2103 | 58 | |
| AK162214.1 | - | 830 | 58 | |||
| BC039282.1 | - | 4 - 5 | 491 | 4976266 | 58 | |
Set Membership: |
|