Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00557967_m1
          See other PP2D1 GE Assays ›
          Gene Symbol:
          Pp2d1
          Gene Name:
          protein phosphatase 2C-like domain containing 1
          Gene Aliases:
          4921523A10Rik
          Chromosome Location:
          Chr.17: 53507460 - 53539451 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00557967_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pp2d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173449.3 NP_775625.1 2 - 3 1338 106
          GenBank mRNA AK076579.1 - 2 - 3 1339 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002282.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pp2d1

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          catalytic activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185569 AK076579.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2185569 NM_173449.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556498 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543116 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 904617 AF045483.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 804666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312318 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 179205 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116883 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152639 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173922 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185569 AK076579.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2185569 NM_173449.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098845 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08806041 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116883 - -
          Whole Mouse Genome G4122A A_51_P456248 BC096481.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456248 AK159706.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456248 BC004842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456248 AK150405.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P456248 AK151066.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107958 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173922 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31244023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31243898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13557033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13529991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13155638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13531565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13486355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13532463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13529988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13529987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31244041 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451801 - -
          Affymetrix Mouse Gene 1.1 ST Array 10451800 - -
          Affymetrix Mouse Clariom S Array PSR1700021383.mm.1 NM_173449.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021383.mm.1 NM_173449.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021383.mm.1 TC1700002282.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 637707 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030253 - -
          Illumina mouse-6 v1.0 expression beadchip 2760138 - -
          Illumina mouse-6 v1.0 expression beadchip 2030253 - -
          Illumina mouse-6 v1.0 expression beadchip 1850066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002760138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641284 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755841 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244138 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641284 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755841 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244138 - -
          mouse-6 v1.1 (Illumina) scl0110332.3_9-S - -
          mouse-6 v1.1 (Illumina) scl00110332.2_7-S - -
          mouse-6 v1.1 (Illumina) scl00110332.2_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 2030253 - -
          Illumina Sentrix mouse-6.v1 bead array 1850066 - -
          Illumina Sentrix mouse-6.v1 bead array 2760138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00110332.2_82-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00110332.2_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0110332.3_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030253 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850066 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2760138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2861482 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755841 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641284 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244138 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755841 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2861482 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244138 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641284 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0