Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00624065_s1
          See other A730013G03RIK GE Assays ›
          Gene Symbol:
          A730013G03Rik
          Gene Name:
          RIKEN cDNA A730013G03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 192833173 - 192835808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00624065_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730013G03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK042646.1 - 1 1157 100
          AK082188.1 - 1 1650 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P127149 AK042646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P127149 AK082188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P127149 AK042646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P127149 AK082188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077831 AK042646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077831 AK082188.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949912 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131109 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077831 AK042646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077831 AK082188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P127149 AK042646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P127149 AK082188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P127149 AK042646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P127149 AK082188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127149 AK042646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127149 AK082188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127149 AK042646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127149 AK082188.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00073150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00128263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00633970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01086521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00128267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00128266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00633964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00050249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00128268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00073138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01086520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20945739 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 515491 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840286 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0