Search
Search

4933409G03Rik| Gene Symbol: 4933409G03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177651.3 | NP_808319.1 | 5 - 6 | 328 | 84 | |
| GenBank mRNA | AK077128.1 | - | 5 - 6 | 329 | 84 | |
Set Membership: |
|