Search
Search

Adam2| Gene Symbol: Adam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009618.2 | NP_033748.2 | 14 - 15 | 1528 | 64 | |
| GenBank mRNA | AK016550.1 | - | 14 - 15 | 1528 | 64 | |
| AK161266.1 | - | 14 - 15 | 1517 | 64 | ||
| U16242.1 | - | 14 - 15 | 1517 | 64 | ||
| U22057.1 | - | 13 - 14 | 1468 | 64 | ||
| U38806.1 | - | 14 - 15 | 1533 | 64 | ||
Set Membership: |
|