Search
Search

Atg4d| Gene Symbol: Atg4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153583.10 | NP_705811.8 | 5 - 6 | 1027 | 90 | |
| GenBank mRNA | AK036282.1 | - | 5 - 6 | 1015 | 90 | |
| AK036571.1 | - | 5 - 6 | 1016 | 90 | ||
| AK081002.1 | - | 5 - 6 | 1014 | 90 | ||
| AK081605.1 | - | 5 - 6 | 1005 | 90 | ||
| AK154130.1 | - | 5 - 6 | 1020 | 90 | ||
| BC030861.1 | - | 5 - 6 | 1004 | 4208700 | 90 | |
| BC069851.1 | - | 5 - 6 | 979 | 6490007 | 90 | |
Set Membership: |
|