| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400003061.mm.1 |
NM_001081221.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003061.mm.1 |
NM_001081221.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003061.mm.1 |
TC1400000347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003064.mm.1 |
NM_001081221.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003064.mm.1 |
NM_001081221.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003064.mm.1 |
TC1400000347.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400003063.mm.1 |
NM_001081221.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003063.mm.1 |
NM_001081221.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400003063.mm.1 |
TC1400000347.mm.1 |
MTA TC |