Search
Search

Klhl36| Gene Symbol: Klhl36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146219.1 | NP_666331.1 | 2 - 3 | 226 | 67 | |
| GenBank mRNA | AK131135.1 | - | 2 - 3 | 151 | 67 | |
| AK157027.1 | - | 2 - 3 | 260 | 67 | ||
| AK164240.1 | - | 2 - 3 | 266 | 67 | ||
| AK164831.1 | - | 2 - 3 | 260 | 67 | ||
| BC025816.1 | - | 2 - 3 | 226 | 5098064 | 67 | |
Set Membership: |
|