Search
Search

Sipa1l2| Gene Symbol: Sipa1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081337.1 | NP_001074806.1 | 16 - 17 | 4743 | 58 | |
| GenBank mRNA | AK028440.1 | - | 16 - 17 | 3855 | 58 | |
| AK122501.1 | - | 14 - 15 | 3020 | 58 | ||
| BC058408.1 | - | 7 - 8 | 1216 | 58 | ||
| BC072593.1 | - | 18 - 19 | 5029 | 58 | ||
| BC089367.1 | - | 6 - 7 | 1129 | 58 | ||
| BC150682.1 | - | 16 - 17 | 4788 | 9087593 | 58 | |
Set Membership: |
|