| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600013498.mm.1 |
NR_027803.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013498.mm.1 |
NR_027803.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013498.mm.1 |
TC0600001697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600013493.mm.1 |
NR_027803.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013493.mm.1 |
NR_027803.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013493.mm.1 |
TC0600001697.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600013494.mm.1 |
NR_027803.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013494.mm.1 |
NR_027803.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600013494.mm.1 |
TC0600001697.mm.1 |
MTA TC |