Search
Search

Tuft1| Gene Symbol: Tuft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293728.1 | NP_001280657.1 | 9 - 10 | 933 | 68 | |
| NM_011656.3 | NP_035786.1 | 10 - 11 | 1008 | 68 | ||
| GenBank mRNA | AF002860.1 | - | 10 - 11 | 902 | 68 | |
| AF047704.1 | - | 10 - 11 | 942 | 68 | ||
| AK132488.1 | - | 10 - 11 | 940 | 68 | ||
| AK171705.1 | - | 9 - 10 | 898 | 68 | ||
| BC019213.1 | - | 10 - 11 | 939 | 5064658 | 68 | |
Set Membership: |
|