Search
Search

Ttc39c| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028341.4 | NP_082617.2 | 3 - 4 | 691 | 92 | |
| GenBank mRNA | AK013267.1 | - | 3 - 4 | 426 | 92 | |
| BC020021.1 | - | 3 - 4 | 441 | 2648772 | 92 | |
Set Membership: |
|