Search
Search

Gm166| Gene Symbol: Gm166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033040.3 | NP_001028212.1 | 4 - 5 | 985 | 67 | |
| GenBank mRNA | AK140121.1 | - | 4 - 5 | 986 | 67 | |
| BC066021.1 | - | 4 - 5 | 566 | 5717506 | 67 | |
Set Membership: |
|