Search
Search

Zfp839| Gene Symbol: Zfp839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199785.1 | NP_001186714.1 | 4 - 5 | 1401 | 60 | |
| NM_028365.2 | NP_082641.1 | 5 - 6 | 1629 | 60 | ||
| GenBank mRNA | AK013356.1 | - | 4 - 5 | 1374 | 60 | |
| BC150732.1 | - | 5 - 6 | 1629 | 9087643 | 60 | |
Set Membership: |
|