Search
Search

Arhgap31| Gene Symbol: Arhgap31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020260.2 | NP_064656.2 | 1 - 2 | 457 | 63 | |
| GenBank mRNA | AF151363.1 | - | 1 - 2 | 98 | 63 | |
| AK018057.1 | - | 1 - 2 | 323 | 63 | ||
| AK045213.1 | - | 1 - 2 | 321 | 63 | ||
| AK138417.1 | - | 1 - 2 | 697 | 63 | ||
| AK149975.1 | - | 1 - 2 | 293 | 63 | ||
| AK150109.1 | - | 1 - 2 | 293 | 63 | ||
| AK155729.1 | - | 1 - 2 | 277 | 63 | ||
| AK170685.1 | - | 1 - 2 | 277 | 63 | ||
| BB865916.1 | - | 1 - 2 | 459 | 63 | ||
| BC138870.1 | - | 1 - 2 | 262 | 8861892 | 63 | |
Set Membership: |
|