| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600010367.mm.1 |
NM_029779.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010367.mm.1 |
NM_029779.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010367.mm.1 |
TC1600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010370.mm.1 |
TC1600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010368.mm.1 |
NM_029779.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010368.mm.1 |
NM_029779.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600010368.mm.1 |
TC1600001281.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600010374.mm.1 |
TC1600001281.mm.1 |
MTA TC |