Search
Search

Eif1b| Gene Symbol: Eif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026892.3 | NP_081168.1 | 2 - 3 | 405 | 90 | |
| GenBank mRNA | AK005192.1 | - | 2 - 3 | 372 | 90 | |
| AK013965.1 | - | 2 - 3 | 378 | 90 | ||
| BC030319.1 | - | 2 - 3 | 274 | 5372299 | 90 | |
| BC033505.1 | - | 2 - 3 | 376 | 4209443 | 90 | |
Set Membership: |
|