| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800018377.mm.1 |
NM_026964.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018377.mm.1 |
NM_026964.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018377.mm.1 |
TC0800002410.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018376.mm.1 |
NM_026964.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018376.mm.1 |
NM_026964.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018376.mm.1 |
TC0800002410.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800018378.mm.1 |
NM_026964.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018378.mm.1 |
NM_026964.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800018378.mm.1 |
TC0800002410.mm.1 |
MTA TC |