Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013580.mm.1 |
NM_001104543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013580.mm.1 |
TC1700001525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013579.mm.1 |
NM_001104543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013579.mm.1 |
TC1700001525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013583.mm.1 |
NM_001104543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013583.mm.1 |
TC1700001525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013581.mm.1 |
NM_001104543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013581.mm.1 |
TC1700001525.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013582.mm.1 |
NM_001104543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013582.mm.1 |
TC1700001525.mm.1 |
MTA TC |