| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200024980.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024980.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024980.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024975.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024975.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024975.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024976.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024976.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024976.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024979.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024979.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024979.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024973.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024973.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024973.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024977.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024977.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024977.mm.1 |
TC0200003027.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200024978.mm.1 |
NM_028317.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024978.mm.1 |
NM_028317.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200024978.mm.1 |
TC0200003027.mm.1 |
MTA TC |