| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600000062.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000062.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000062.mm.1 |
TC1600000014.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000064.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000064.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000064.mm.1 |
TC1600000014.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000065.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000065.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000065.mm.1 |
TC1600000014.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000063.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000063.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000063.mm.1 |
TC1600000014.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000061.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000061.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000061.mm.1 |
TC1600000014.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600000066.mm.1 |
NM_001081217.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000066.mm.1 |
NM_001081217.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600000066.mm.1 |
TC1600000014.mm.1 |
MTA TC |