| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00019781.mm.1 |
NM_001164686.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019781.mm.1 |
NM_001164686.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019781.mm.1 |
TC0X00003159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019782.mm.1 |
NM_001164686.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019782.mm.1 |
NM_001164686.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019782.mm.1 |
TC0X00003159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019783.mm.1 |
NM_001164686.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019783.mm.1 |
NM_001164686.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019783.mm.1 |
TC0X00003159.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00019780.mm.1 |
NM_001164686.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019780.mm.1 |
NM_001164686.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00019780.mm.1 |
TC0X00003159.mm.1 |
MTA TC |