| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700013602.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013602.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013602.mm.1 |
TC1700001534.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013604.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013604.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013604.mm.1 |
TC1700001534.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013605.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013605.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013605.mm.1 |
TC1700001534.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013607.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013607.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013607.mm.1 |
TC1700001534.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013603.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013603.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013603.mm.1 |
TC1700001534.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700013606.mm.1 |
NM_001104567.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013606.mm.1 |
NM_001104567.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700013606.mm.1 |
TC1700001534.mm.1 |
MTA TC |