Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033596.3 | NP_291074.1 | 1 | 49 | 84 | |
GenBank mRNA | BC051566.1 | - | 1 | 57 | 84 | |
BC057955.1 | - | 43 | 5359141 | 84 | ||
BX520915.1 | - | 1 | 49 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |