Search
Search

Mir34b| Gene Symbol: Mir34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029655.1 | 1 | 44 | 67 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 34c | Mir34c |
Mirn34c, mir-34c, mmu-mir-34c |
-452.0 | Sense | Chr. 9 - 51103034 - 51103110 on NCBI Build GRCm38 |
| Downstream |
POU domain, class 2, associating factor 1 |
Pou2af1 |
BOB.1, Bob-1, Bob1, OBF-1, OBF.1, OCA-B, OCAB |
110.0kb | Anti-sense | Chr. 9 - 51213690 - 51240079 on NCBI Build GRCm38 |
Set Membership: |
|