Search
Search

BC048507| Gene Symbol: BC048507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001185.3 | NP_001001185.1 | 1 | 418 | 92 | |
| GenBank mRNA | AK133011.1 | - | 1 | 419 | 92 | |
| BC048507.1 | - | 1 | 407 | 6774067 | 92 | |
Set Membership: |
|