Search
Search

Supt4a| Gene Symbol: Supt4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009296.1 | NP_033322.1 | 5 | 520 | 60 | |
| GenBank mRNA | AK002990.1 | - | 5 | 622 | 60 | |
| AK146235.1 | - | 6 | 677 | 60 | ||
| AK167941.1 | - | 5 | 534 | 60 | ||
| BC024391.1 | - | 5 | 515 | 5011064 | 60 | |
| BC061174.1 | - | 5 | 521 | 6592996 | 60 | |
| BC087923.1 | - | 5 | 496 | 30308013 | 60 | |
| U43154.1 | - | 5 | 486 | 60 | ||
Set Membership: |
|