Search
Search

C13H1orf146| Gene Symbol: C13H1orf146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017346089.1 | XP_017201578.1 | 5 - 6 | 1383 | 93 | |
| XM_017346090.1 | XP_017201579.1 | 5 - 6 | 1383 | 93 | ||
| XM_017346091.1 | XP_017201580.1 | 5 - 6 | 1275 | 93 | ||
Set Membership: |
|