Search
Search

TAZ| Gene Symbol: TAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032914.1 | NP_001028086.1 | 290 | 66 | ||
| XM_015128467.1 | XP_014983953.1 | 760 | 66 | |||
| XM_015128468.1 | XP_014983954.1 | 760 | 66 | |||
| XM_028841631.1 | XP_028697464.1 | 760 | 66 | |||
| XM_028841632.1 | XP_028697465.1 | 760 | 66 | |||
| XM_028841633.1 | XP_028697466.1 | 210 | 66 | |||
| XM_028841635.1 | XP_028697468.1 | 760 | 66 | |||
| GenBank mRNA | AY621059.1 | - | 290 | 66 | ||
| JU492311.1 | - | 535 | 66 | |||
Set Membership: |
|