Search
Search

RAB28| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265724.1 | NP_001252653.1 | 394 | 132 | ||
| NM_001376264.1 | NP_001363193.1 | 477 | 132 | |||
| XM_015138003.2 | XP_014993489.1 | 1055 | 132 | |||
| GenBank mRNA | JU321729.1 | - | 438 | 132 | ||
| JU477881.1 | - | 378 | 132 | |||
| JV048728.1 | - | 371 | 132 | |||
| JV600587.1 | - | 464 | 132 | |||
| JV635599.1 | - | 394 | 132 | |||
Set Membership: |
|