Search
Search
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108304.1 | NP_001101774.1 | 13 - 14 | 2644 | 81 | |
| XM_006247719.2 | XP_006247781.1 | 14 - 15 | 1830 | 81 | ||
| XM_006247720.2 | XP_006247782.1 | 12 - 13 | 1968 | 81 | ||
| XM_006247721.1 | XP_006247783.1 | 13 - 14 | 1654 | 81 | ||
Set Membership: |
|