Search
Search
| Gene Symbol: Kcnj13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053608.2 | NP_446060.1 | 2 - 3 | 573 | 105 | |
| XM_008767301.1 | XP_008765523.1 | 1 - 2 | 1908 | 105 | ||
| GenBank mRNA | AJ006129.1 | - | 1 - 2 | 456 | 105 | |
Set Membership: |
|