Search
Search

Dgcr2
LOC100910275| Gene Symbol: Dgcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012146.1 | NP_001012146.1 | 1 - 2 | 362 | 79 | |
| XM_008768890.1 | XP_008767112.1 | 1 - 2 | 336 | 79 | ||
| GenBank mRNA | BC081905.1 | - | 1 - 2 | 362 | 7111034 | 79 |
| FQ211720.1 | - | 1 - 2 | 286 | 79 | ||
| FQ228410.1 | - | 1 - 2 | 264 | 79 | ||
| Gene Symbol: LOC100910275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750241.2 | XP_003750289.1 | 1 - 2 | 361 | 79 | |
| XM_006240675.2 | XP_006240737.1 | 1 - 2 | 329 | 79 | ||
Set Membership: |
|